ENA258389853 Enamine

7-methoxy-4-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD09730079
  • Cas Number : 72985-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60025
Availability details for
Quantity
  • Availability
  • Add to Cart