ENA258389857 Enamine

4-methyl-5-(methylsulfanyl)-1,3-thiazol-2-amine

  • MDL number MFCD09938994
  • Cas Number : 34006-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60030
Availability details for
Quantity
  • Availability
  • Add to Cart