ENA258389863 Enamine

2-(2,2-dimethylpropyl)pyrrolidine

  • MDL number MFCD02663457
  • Cas Number : 383127-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60048
Availability details for
Quantity
  • Availability
  • Add to Cart