ENA258389864 Enamine

hexahydro-1H-pyrrolo[2,1-c]morpholin-3-ylmethanamine

  • MDL number MFCD11934396
  • Cas Number : 1181643-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60049
Availability details for
Quantity
  • Availability
  • Add to Cart