ENA258389866 Enamine

1-(2,2,2-trifluoroethyl)pyrrolidin-3-amine

  • MDL number MFCD11640761
  • Cas Number : 1096861-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60051

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-60051
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart