ENA258389873 Enamine

2-fluoropyridine-4-carboxamide

  • MDL number MFCD14618467
  • Cas Number : 369-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-174
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60064
Availability details for
Quantity
  • Availability
  • Add to Cart