ENA258389875 Enamine

2-[(1H-indazol-6-yl)formamido]propanoic acid

  • MDL number MFCD11648627
  • Cas Number : 1397007-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60068
Availability details for
Quantity
  • Availability
  • Add to Cart