ENA258389877 Enamine

6-amino-5-chloropyridine-3-carboxylic acid

  • MDL number MFCD11132184
  • Cas Number : 41668-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60072
Availability details for
Quantity
  • Availability
  • Add to Cart