ENA258389879 Enamine

3-(chloromethyl)-5-[(3,4-dimethoxyphenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD11541537
  • Cas Number : 1038981-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-55492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55492
Availability details for
Quantity
  • Availability
  • Add to Cart