ENA258389882 Enamine

6-amino-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD01346678
  • Cas Number : 18802-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 334...336
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60078

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60078
Availability details for
Quantity
  • Availability
  • Add to Cart