ENA258389887 Enamine

(2,6-dichlorophenyl)methanethiol

  • MDL number MFCD10691744
  • Cas Number : 66279-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60083

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60083
Availability details for
Quantity
  • Availability
  • Add to Cart