ENA258389890 Enamine

4-[(2-methoxyethyl)amino]benzonitrile

  • MDL number MFCD11135500
  • Cas Number : 1019600-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60086

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60086
Availability details for
Quantity
  • Availability
  • Add to Cart