ENA258389891 Enamine

2-methyl-2-phenylpropan-1-ol

  • MDL number MFCD00506139
  • Cas Number : 2173-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60087
Availability details for
Quantity
  • Availability
  • Add to Cart