ENA258389898 Enamine

1-chloro-2-(2-chloroethyl)benzene

  • MDL number MFCD11615165
  • Cas Number : 1005-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60096
Availability details for
Quantity
  • Availability
  • Add to Cart