ENA258389902 Enamine

1-cyclopropyl-1,4-diazepane

  • MDL number MFCD14650978
  • Cas Number : 30858-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60101

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60101
Availability details for
Quantity
  • Availability
  • Add to Cart