ENA258389905 Enamine

3-(difluoromethoxy)benzaldehyde

  • MDL number MFCD00236222
  • Cas Number : 85684-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60104

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60104
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart