ENA258389906 Enamine

6-(difluoromethoxy)-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD14705868
  • Cas Number : 1221723-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60105
Availability details for
Quantity
  • Availability
  • Add to Cart