ENA258389914 Enamine

5-(thiophen-2-yl)-1,2-oxazole-3-carboxamide

  • MDL number MFCD11643942
  • Cas Number : 1019656-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60119
Availability details for
Quantity
  • Availability
  • Add to Cart