ENA258389917 Enamine

3-chloro-5-cyclopropyl-8-thia-4,6-diazatricyclo[7.4.0.0^{2,7}]trideca-1(9),2,4,6-tetraene

  • MDL number MFCD03124472
  • Cas Number : 462066-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112...114
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60122

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60122
Availability details for
Quantity
  • Availability
  • Add to Cart