ENA258389921 Enamine

{1-[(thiophen-2-yl)methyl]piperidin-4-yl}methanamine

  • MDL number MFCD06801314
  • Cas Number : 883541-34-2
  • Molecular Weight : 210.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60126
Availability details for
Quantity
  • Availability
  • Add to Cart