ENA258389924 Enamine

(1-bromopropyl)benzene

  • MDL number MFCD00055549
  • Cas Number : 2114-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60130

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60130
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart