ENA258389926 Enamine

7-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD11574543
  • Cas Number : 17349-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60132

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60132
Availability details for
Quantity
  • Availability
  • Add to Cart