ENA258389927 Enamine

6-(3-nitrophenyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD08062116
  • Cas Number : 54558-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274...276
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60133
Availability details for
Quantity
  • Availability
  • Add to Cart