ENA258389932 Enamine

4-[(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-N'-hydroxybenzene-1-carboximidamide

  • Cas Number : 1158102-09-0
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60165
Availability details for
Quantity
  • Availability
  • Add to Cart