ENA258389943 Enamine

N'-hydroxy-4-phenylbutanimidamide

  • MDL number MFCD09808080
  • Cas Number : 175532-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60182

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60182
Availability details for
Quantity
  • Availability
  • Add to Cart