ENA258389946 Enamine

2-amino-2-methylbutan-1-ol

  • MDL number MFCD09759242
  • Cas Number : 10196-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60187
Availability details for
Quantity
  • Availability
  • Add to Cart