ENA258389948 Enamine

3-methyl-6-(pyridin-4-yl)pyrido[3,2-d][1,2]oxazole-4-carboxylic acid

  • MDL number MFCD09739881
  • Cas Number : 954262-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56158

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56158
Availability details for
Quantity
  • Availability
  • Add to Cart