ENA258389953 Enamine

diphenyl-1,2-oxazol-5-amine

  • MDL number MFCD03861189
  • Cas Number : 52392-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60202
Availability details for
Quantity
  • Availability
  • Add to Cart