ENA258389955 Enamine

[4-(2,3-dihydro-1-benzofuran-5-yl)phenyl]methanamine

  • MDL number MFCD12861457
  • Cas Number : 1183374-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60224
Availability details for
Quantity
  • Availability
  • Add to Cart