ENA258389959 Enamine

3-(2-nitrophenyl)propanoic acid

  • MDL number MFCD00130002
  • Cas Number : 2001-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56341
Availability details for
Quantity
  • Availability
  • Add to Cart