ENA258389960 Enamine

5-cyclopropyl-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11118807
  • Cas Number : 1092394-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60233

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60233
Availability details for
Quantity
  • Availability
  • Add to Cart