ENA258389966 Enamine

5-(2,5-dimethoxyphenyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD07377123
  • Cas Number : 953752-64-2
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2,5-dimethoxyphenyl)-3-isoxazolecarboxylic acid
Inchi Code 1S/C12H11NO5/c1-16-7-3-4-10(17-2)8(5-7)11-6-9(12(14)15)13-18-11/h3-6H,1-2H3,(H,14,15)
Inchi Key SHKJDWPURNWNTI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60245

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-60245
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart