ENA258389968 Enamine

1-[(quinolin-8-yl)methyl]-1H-pyrazol-3-amine

  • MDL number MFCD12719944
  • Cas Number : 1183577-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60250
Availability details for
Quantity
  • Availability
  • Add to Cart