ENA258389972 Enamine

ethyl 2-(6-amino-1H-indazol-1-yl)acetate

  • MDL number MFCD09727484
  • Cas Number : 624720-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60257

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60257
Availability details for
Quantity
  • Availability
  • Add to Cart