ENA258389973 Enamine

1-(4-hydroxypiperidin-1-yl)butan-1-one

  • MDL number MFCD11044216
  • Cas Number : 202647-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60259
Availability details for
Quantity
  • Availability
  • Add to Cart