ENA258389985 Enamine

4-[2-(thiomorpholin-4-yl)ethyl]aniline

  • MDL number MFCD11172340
  • Cas Number : 1042786-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60298
Availability details for
Quantity
  • Availability
  • Add to Cart