ENA258389986 Enamine

2-bromo-1-(2,3-dihydro-1H-inden-5-yl)ethan-1-one

  • MDL number MFCD09745882
  • Cas Number : 39696-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60302

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60302
Availability details for
Quantity
  • Availability
  • Add to Cart