ENA258389987 Enamine

2-phenylbenzene-1-sulfonamide

  • MDL number MFCD11650103
  • Cas Number : 40182-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-119
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60307
Availability details for
Quantity
  • Availability
  • Add to Cart