ENA258389993 Enamine

methyl 4-cyanobutanoate

  • MDL number MFCD12168338
  • Cas Number : 41126-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60318

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60318
Availability details for
Quantity
  • Availability
  • Add to Cart