ENA258389994 Enamine

6-methyl-3,4-dihydro-2H-1-benzopyran

  • MDL number MFCD14705883
  • Cas Number : 3722-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60319
Availability details for
Quantity
  • Availability
  • Add to Cart