ENA258389995 Enamine

4-bromo-2-chloro-1-(chloromethyl)benzene

  • MDL number MFCD12065644
  • Cas Number : 185315-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60320
Availability details for
Quantity
  • Availability
  • Add to Cart