ENA258389996 Enamine

N-methyl-1-(4-methylcyclohexyl)piperidin-4-amine

  • MDL number MFCD11640986
  • Cas Number : 1096836-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60321
Availability details for
Quantity
  • Availability
  • Add to Cart