ENA258389999 Enamine

2-(4-methylphenyl)-2-(pyrrolidin-1-yl)ethan-1-amine

  • MDL number MFCD08445808
  • Cas Number : 889940-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60324
Availability details for
Quantity
  • Availability
  • Add to Cart