ENA258390001 Enamine

propyl 2-amino-5-bromobenzoate

  • MDL number MFCD12149109
  • Cas Number : 1178634-57-5
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60326
Availability details for
Quantity
  • Availability
  • Add to Cart