ENA258390003 Enamine

2-ethylmorpholine hydrochloride

  • MDL number MFCD14705612
  • Cas Number : 1221722-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56464
Availability details for
Quantity
  • Availability
  • Add to Cart