ENA258390014 Enamine

2-cyano-3-[4-(dimethylamino)phenyl]prop-2-enamide

  • MDL number MFCD00025491
  • Cas Number : 3785-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15632
Availability details for
Quantity
  • Availability
  • Add to Cart