ENA258390033 Enamine

3,6-dichloropyridine-2-carbonitrile

  • MDL number MFCD00546824
  • Cas Number : 1702-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57209
Availability details for
Quantity
  • Availability
  • Add to Cart