ENA258390081 Enamine

1-[1-(4-fluorophenyl)-5-methyl-1H-pyrazol-4-yl]ethan-1-amine

  • MDL number MFCD13621149
  • Cas Number : 1152949-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57724

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57724
Availability details for
Quantity
  • Availability
  • Add to Cart