ENA258390094 Enamine

4-[(2-methoxy-5-methylphenyl)amino]butanoic acid

  • MDL number MFCD09808200
  • Cas Number : 1016735-33-3
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58562
Availability details for
Quantity
  • Availability
  • Add to Cart