ENA258390097 Enamine

1,3-diphenylpropan-2-amine

  • MDL number MFCD00449548
  • Cas Number : 4275-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33167
Availability details for
Quantity
  • Availability
  • Add to Cart